Learn More
Hach Company Aluminum TNTplus Vial Test (0.02-0.50 mg/L Al), 24 Tests
Supplier:  Hach Company TNT848
Encompass
Aluminum TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination Aluminum by the Chromazurol S Method.
Catalog No. 50-199-8139