Learn More
Apexbio Technology LLC Alverine Citrate 5560-59-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B16555.1
Encompass_Preferred
Alverine citrate (CAS 5560-59-8) is a synthetic small-molecule compound employed primarily in studies of functional gastrointestinal disorders It exerts antispasmodic effects predominantly via calcium channel blockade reducing smooth muscle contraction and alleviating gastrointestinal symptoms Biomedical researchers utilize alverine citrate to examine gut motility mechanisms visceral hypersensitivity pathways and potential therapeutic approaches for gastrointestinal dysfunction
Catalog No. 50-000-63760