Learn More
Selleck Chemical LLC Alverine Citrate S3054-25mg
SDP
Supplier:  Selleck Chemical LLC S305425MG
Encompass_Preferred
Alverine citrate (NSC 35459) is a drug used for functional gastrointestinal disorders Alverine citrate is a 5-HT1A receptor antagonist with an IC50 of 101 nM
Catalog No. 50-311-1596