Learn More
Medchemexpress LLC Alverine citrate | 5560-59-8 | MFCD00035086 | 99.5% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB050010G
Encompass_Preferred
Alverine citrate is a 5-HT1A receptor antagonist, inhibiting the receptor with an IC50 of 101 nM. It is utilized in the treatment of functional gastrointestinal disorders, acting directly on gut muscles to induce relaxation. In vitro studies demonstrate its high affinity for 5-HT1A receptors and weak affinity for 5-HT3 and 5-HT4 subtypes. This compound is suitable for research applications.
  • 5-HT1A receptor antagonist with an IC50 of 101 nM
  • Used for functional gastrointestinal disorders
  • Acts directly on gut muscle to cause relaxation
  • High affinity for 5-HT1A receptors
  • Weak affinity for 5-HT3 and 5-HT4 subtypes
  • May increase Ca influx and suppress evoked activity
  • For research use only
Catalog No. 50-004-09486