Learn More
Selleck Chemical LLC Alvespimycin (17-DMAG) HCl 10mM/1mL 467214-21-7 NSC 707545,BMS 826476 HCl,KOS 1022
SDP
Supplier:  Selleck Chemical LLC S114210MM/1ML
Encompass_Preferred
Alvespimycin (17-DMAG, NSC 707545, BMS 826476, KOS 1022) HCl is a potent HSP90 inhibitor with IC50 of 62 nM in a cell-free assay. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0951