Learn More
TARGETMOL CHEMICALS INC ALVESPIMYCIN HYDROCHLORI 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T629750MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alvespimycin hydrochloride (BMS 826476) is a potent HSP90 inhibitor with IC50 of 62 nM. Phase 2. purity: 99%
Catalog No. 50-257-9449