Learn More
Medchemexpress LLC Alvespimycin hydrochloride | 467214-21-7 | 98.8% | 1 ML

Supplier: Medchemexpress LLC HY1202410MM 1ML
Alvespimycin hydrochloride is the hydrochloride salt of 17-DMAG, a potent small-molecule inhibitor of heat shock protein 90 (Hsp90) used in biochemical and cell-based research to probe protein homeostasis and apoptosis pathways. It is supplied for laboratory research in solution or solid form and should be handled according to safety guidance.
- Potent Hsp90 binding activity (EC50 ≈ 62 ± 29 nM).
- Available as 10 mM solution in DMSO (1 mL) or as solid in multiple mass sizes.
- High purity (≈98.8%).
- Suitable for biochemical and cell-based assays.
- Storage: 4°C sealed; in solvent -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.