Learn More
Selleck Chemical LLC AM 095 5mg
SDP
Supplier:  Selleck Chemical LLC S66335MG
Encompass_Preferred
AM095 is an antagonist of the LPA type 1 receptor with IC50 values of 0.98 and 0.73 ?M for recombinant human and mouse LPA1, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3193