Learn More
Selleck Chemical LLC AM1241 25mg 444912-48-5
SDP
Supplier:  Selleck Chemical LLC S154425MG
Encompass_Preferred
AM-1241 is a selective cannabinoid CB2 receptor agonist with Ki of 3.4 nM, exhibits 82-fold selectivity over CB1 receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-770-1