Learn More
Enzo Life Sciences AM-251 (10mg). CAS: 183232-66-8
SDP
Supplier:  Enzo Life Sciences BMLCR1080010
Encompass_Preferred
Cannabinoid CB1 receptor antagonist (Ki=7.5 nM). Displays a high level of selectivity (306-fold) for CB1 over CB2 receptors. Structurally related to the cannabinoid receptor (CB) antagonist SR 141716A. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, 100% ethanol or dimethyl formamide. Long Term Storage: +4°C.
Catalog No. 50-201-0472