Learn More
Cayman Chemical AM1172 Fatty Acids and Derivat
SDP
Supplier:  Cayman Chemical 100052235 MG
Encompass_Preferred
A hydrolysis-resistant endocannabinoid analog that inhibits AEA cellular uptake; resistant to FAAH hydrolysis; is the structurally “reversed” isomer of AM404; blocks the uptake of tritiated AEA (EC50 = ~1.5 µM in mouse cortical neurons)
Catalog No. 50-288-8788