Learn More
Apexbio Technology LLC AM251, 5mg. Cas: 183232-66-8 MFCD: MFCD01861181. Potent CB1 antagonist
SDP
Supplier:  Apexbio Technology LLC B14275
Encompass_Preferred
AM521 is a potent cannabinoid 1 (CB1) receptor antagonist with IC50 of 8 nM and Ki of 7.49 nM.The cannabinoid receptor (CB1 and CB2) is a member of G-protein coupled receptor which plays a significant role in physiologic processes such as cognitive and immune functions. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5234