Learn More
Dino Lite HDMI Digital Microscope AM5218MZTL- 720p, 5x - 140x Optical Magnification, Polarized Light, Long Working Distance
Supplier:  Dino Lite AM5218MZTL
Encompass_Preferred
The Dino-Lite Edge Series AM5218MZTL is designed for connecting directly to HDTV or LCD monitors. The connection uses DVI or HD interface with HD 720p format. AM5218MZTL offers a full magnification range of 10x - 140x. Polarization can be switched on/off or can be adjusted to offer full or half polarization for controlling glare. The AM5218MZTL's MicroTouch provides the convenience to freeze the live video to examine details by pressing and holding the MicroTouch sensor for 1 second. The LEDs can also be switched by holding the same MicroTouch sensor for two seconds or more.
Catalog No. NC1614662