Learn More
Cayman Chemical AM694 NpentnoIc ACD metao 10mg
SDP
Supplier:  Cayman Chemical 1435910 MG
Encompass_Preferred
An expected urinary metabolite of AM694, based on the known metabolism of similar compounds; intended for forensic and research applications
Catalog No. 50-295-2844