Learn More
Medchemexpress LLC Amantadine hydrochloride | 665-66-7 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB0402A5G
Encompass_Preferred
Amantadine hydrochloride is an orally active and potent antiviral agent, effective against influenza A viruses. It inhibits various ion channels, including NMDA, M2, and Coronavirus ion channels. The compound also demonstrates anti-orthopoxvirus and anticancer activities, and is utilized in research for Parkinson's disease, postoperative cognitive dysfunction (POCD), and COVID-19.
  • Potent antiviral agent, effective against influenza A viruses and SARS-CoV-2 replication
  • Inhibits various ion channels, including NMDA, M2, and Coronavirus
  • Exhibits anti-orthopoxvirus and anticancer activities
  • Used in research for Parkinson's disease, postoperative cognitive dysfunction, and COVID-19
  • Induces apoptosis and arrests cell cycle in cancer cells
  • Inhibits surgery-induced neuroinflammation and learning/memory impairment
Catalog No. 50-004-08580