Learn More
Medchemexpress LLC HY-P1576A 1mg Medchemexpress, AMARA peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1576A1MG
Encompass_Preferred
Medchemexpress, HY-P1576A 1mg AMARA peptide (TFA) CAS: AMARA peptide (TFA) is a substrate for salt-inducible kinase (SIK) and adenosine monophosphate activated protein kinase (AMPK). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4228