Learn More
Medchemexpress LLC Amas (N-α-maleimidoacet-oxysuccinimide ester) | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12892550MG
Encompass_Preferred
AMAS (N-α-maleimidoacet-oxysuccinimide ester) is a short, non-cleavable heterobifunctional crosslinker containing an N-hydroxysuccinimide (NHS) ester and a maleimide group. It enables covalent conjugation between primary amines and sulfhydryl groups for bioconjugation of proteins, peptides, and other biomolecules. Supplied as a 50 mg package.
  • Reactive groups: NHS ester (amine-reactive) and maleimide (sulfhydryl-reactive).
  • Cleavability: non-cleavable crosslinker.
  • Spacer arm: short spacer for minimal distance between conjugated partners.
  • Applications: amine-to-sulfhydryl conjugation for protein and peptide labeling.
  • Packaging: supplied as a 50 mg vial for laboratory use.
Catalog No. 50-002-03179