Learn More
eMolecules​ 1065267-14-2 | (3,5-Difluoropyridin-2-yl)methanol | Ambeed | MFCD11520700 | 145.109 | C6H5F2NO | 98.000 | OCc1ncc(F)cc1F | 250mg | 494129506
Supplier:  eMolecules​ 494129506
Encompass_Preferred
(3,5-Difluoropyridin-2-yl)methanol | Ambeed | 1065267-14-2 | MFCD11520700 | 145.109 | C6H5F2NO | 98.000 | OCc1ncc(F)cc1F | 250mg | 494129506
Catalog No. 50-001-28120