Learn More
eMolecules​ 171856-09-0 | Fmoc-N-(4-Boc-aminobutyl)-Gly-OH | Ambeed | MFCD04112689 | 468.550 | C26H32N2O6 | 98.000 | CC(C)(C)OC(=O)NCCCCN(CC(O)=O)C(=O)OCC1c2ccccc2-c2ccccc12 | 100mg | 879142932
Supplier:  eMolecules​ 879142932
Encompass_Preferred
Fmoc-N-(4-Boc-aminobutyl)-Gly-OH | Ambeed | 171856-09-0 | MFCD04112689 | 468.550 | C26H32N2O6 | 98.000 | CC(C)(C)OC(=O)NCCCCN(CC(O)=O)C(=O)OCC1c2ccccc2-c2ccccc12 | 100mg | 879142932
Catalog No. 50-001-29763