Learn More
eMolecules​ 97322-87-7 | Troglitazone | Ambeed | MFCD00878416 | 441.540 | C24H27NO5S | 98.000 | Cc1c(C)c2OC(C)(COc3ccc(CC4SC(=O)NC4=O)cc3)CCc2c(C)c1O | 250mg | 528952867
Supplier:  eMolecules​ 528952867
Encompass_Preferred
Troglitazone | Ambeed | 97322-87-7 | MFCD00878416 | 441.540 | C24H27NO5S | 98.000 | Cc1c(C)c2OC(C)(COc3ccc(CC4SC(=O)NC4=O)cc3)CCc2c(C)c1O | 250mg | 528952867
Catalog No. 50-001-29789