Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ambroxide | 6790-58-5 | 236.39 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYN138450MG

Encompass_Preferred

Ambroxide is a naturally occurring terpenoid and a key constituent of ambergris. It is used in the manufacture of high-end perfumes to improve aroma quality and fragrance lasting time.

  • Molecular weight: 236.39
  • Formula: C16H28O
  • CAS No.: 6790-58-5
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, other terpenoids
  • Initial source: Plants (other families), animals (Physeter catodon L.)
  • Purity: 98.0%
  • Solubility in vitro: Soluble in DMSO (25 mg/mL)

Catalog No. 50-004-14793


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.