Learn More
Selleck Chemical LLC Ambroxol HCl 50mg 23828-92-4
SDP
Supplier:  Selleck Chemical LLC S306450MG
Encompass_Preferred
AmbroxolHCl is a potent inhibitor of the neuronal Na+ channels, inhibits TTX-resistant Na+ currents with IC50 of 35.2 ?M and 22.5 ?M for tonic and phasic block, inhibits TTX-sensitive Na+ currents with IC50 of 100 ?M. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3346