Learn More
Cayman Chemical AMCA N-succInImIdyl ester 10mg
SDP
Supplier:  Cayman Chemical 3466810 MG
Encompass_Preferred
An amine-reactive fluorescent protein-labeling agent; abs/em = UV/400-460 nm, respectively; contains an N-hydroxysuccinimide ester that reacts with lysine residues to form photostable amide links; has commonly been used in multiplex immunophenotyping applications
Catalog No. 50-295-0718