Learn More
AdipoGen AMCA-X
SDP
Supplier:  AdipoGen CDXA0073M050
Encompass_Preferred
Chemical. CAS 205124-69-2. Formula C18H22N2O5. MW 346.38. Synthetic. Amine-reactive blue fluorescent probe.
Catalog No. 50-256-1840