Learn More
Medchemexpress LLC AMCA-X SE | 216309-02-3 | ≥97.0% | 443.45 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD10851MG
Encompass_Preferred
AMCA-X-SE is a coumarin derivative that generates fixed blue fluorescence and an NHS-activated ester that forms stable amide bonds with primary amine groups. It is used as a reactive dye for labeling amino groups of peptides, proteins, and oligonucleotides. Maximum excitation/emission wavelength: 354/442 nm.
  • Generates fixed blue fluorescence
  • Forms stable amide bonds with primary amine groups
  • Used as a reactive dye for labeling amino groups
  • Suitable for peptides, proteins, and oligonucleotides
Catalog No. 50-004-28274