Learn More
Selleck Chemical LLC Amcenestrant S9609-25MG
SDP
Supplier:  Selleck Chemical LLC S960925MG
Encompass_Preferred
Amcenestrant (SAR439859 compound 43d) is an orally available and nonsteroidal selective estrogen receptor degrader (SERD) with potential antineoplastic activity SAR439859 is a potent estrogen receptor (ER) antagonist with EC50 of 0 2 nM for ER degradation
Catalog No. 50-000-23983