Learn More
Selleck Chemical LLC Amcinonide
SDP
Supplier:  Selleck Chemical LLC S567125MG
Encompass_Preferred
Amcinonide (Cyclocort CL-34699) is an inhibitor of nitric oxide (NO) release from activated microglia with IC50 of 3 38 nM
Catalog No. 50-000-03075