Learn More
Abcam AMD3100 octahydrochloride, C x CR4 antagonist, 10MG
Supplier:  Abcam AB12071810MG
Encompass_Preferred
MW 794.5 Da, Purity >99%. Plerixafor (hydrochloride) is a macrocyclic compound that acts as an irreversible antagonist against the binding of CXCR4 with its ligand, SDF-1 (CXCL12). It suppresses infection by HIV with an IC₅₀ value of 1-10 ng/ml with selectivity toward CXCR4-tropic virus. Plerixafor mobilizes hematopoietic stem and progenitor cells for transplant better than G-CSF alone. It also increases T-cell trafficking in the blood and spleen as well as the central nervous system. Plerixafor regulates the growth of primary and metastic breast cancer cells and inhibits dissemination of ovarian carcinoma cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4576