Learn More
Medchemexpress LLC Amelenodor | 2389235-01-0 | >98.0% | 399.44 g/mol | C24H21N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1415215MG
Encompass_Preferred
Amelenodor (NX-13) is a small-molecule NLRX1 activator used in research on NOD-like receptor signaling and inflammatory pathways. The compound has the chemical formula C24H21N3O3 and molecular weight 399.44 g/mol, and is supplied in milligram quantities for in vitro and in vivo experimental use.
  • NLRX1 activator for studies of innate immunity and inflammation.
  • High purity suitable for research (>98%).
  • Supplied in small-mass quantities appropriate for dosing and assay preparation.
  • Stability data available for refrigerated and frozen storage conditions.
  • Supporting documentation includes a data sheet and safety data sheet for handling guidance.
Catalog No. 50-002-23288