Learn More
Medchemexpress LLC AMELX Human His-SU 50ug
SDP
Supplier:  Medchemexpress LLC HYP7162750UG
Encompass_Preferred
AMELX Protein plays a central role in biomineralization shaping crystallite formation during the secretory stage of tooth enamel development Its pivotal function extends to structurally organizing and mineralizing developing enamel emphasizing its significance in dental tissue architecture Interactions with KRT5 suggest its involvement in molecular networks contributing to the orchestrated development and mineralization of tooth enamel AMELX Protein Human (His-SUMO) is the recombinant human-derived AMELX protein expressed by E coli with N-SUMO N-6 His labeled tag
Catalog No. 50-001-95430