Learn More
Medchemexpress LLC Amenamevir | 841301-32-4 | 99.9% | 482.55 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14809
Encompass_Preferred
Amenamevir is a potent helicase-primase inhibitor with significant antiviral activity against Herpes Simplex Viruses (HSVs). It effectively inhibits the replication of HSV-1 and HSV-2 strains, demonstrating lower EC50 values compared to acyclovir in both in vitro and in vivo models. This compound is suitable for various laboratory research applications.
  • Potent helicase-primase inhibitor
  • Exhibits antiviral activity against HSVs
  • Inhibits HSV-1 and HSV-2 replication
  • Demonstrates lower EC50 values compared to acyclovir
  • Suitable for in vitro and in vivo research applications
Catalog No. 50-003-62683