Learn More
Medchemexpress LLC Amentoflavone | 1617-53-4 | 538.46 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYN066210MM/1ML
Encompass_Preferred
Amentoflavone is a potent and orally active GABA(A) negative modulator. It exhibits a wide range of biological activities including anti-inflammatory, antioxidative, anti-viral, anti-tumor, anti-radiation, anti-fungal, and antibacterial effects. It also induces apoptosis and cell cycle arrest at the sub-G1 phase.
  • Potent and orally active GABA(A) negative modulator
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidative activity
  • Exhibits anti-viral activity
  • Exhibits anti-tumor activity
  • Exhibits anti-radiation activity
  • Exhibits anti-fungal activity
  • Exhibits antibacterial activity
  • Induces apoptosis
  • Induces cell cycle arrest at the sub-G1 phase
Catalog No. 50-003-88681