Learn More
Med Vet International Ameriod constrictors 5mm, J0659A
Supplier:  Med Vet International JOR659A
Encompass_Preferred
For the surgical treatment of porto caval or hepatic shunts. This special designed product is used to encircle a blood vessel. A special material (hygroscopic compressed casein) inside the donut shaped constrictor imbibes fluid over time. It slowly occludes the blood vessel. A small key is used to lock it in place after it is slipped over the blood vessel. Sizing: about a 50% reduction in the blood vessel diameter should be achieved upon initial surgical placement for the best results. Sterile.
Catalog No. 50-005-84572