Learn More
Medchemexpress LLC Amfenac-d5 sodium | 61941-56-8 | 98.9% | 282.28 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY17479BS1MG
Encompass_Preferred
Amfenac-d5 sodium is a deuterated-labeled form of Amfenac sodium. Amfenac Sodium is an orally active and potent anti-inflammatory compound that possesses antipyretic and analgesic properties. It can be utilized for research into acute and chronic inflammation. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is used as a tracer for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterated-labeled compound.
  • Orally active.
  • Potent anti-inflammatory properties.
  • Antipyretic properties.
  • Analgesic properties.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-86894