Learn More
Medchemexpress LLC Amfenac-d5 sodium | 61941-56-8 | MFCD09842317 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17479BS10MG
Encompass_Preferred
Amfenac-d5 sodium is the deuterium-labeled analog of amfenac sodium supplied as a research-grade stable isotope reference for analytical and pharmacokinetic studies. It is intended for use as an internal standard in mass spectrometry and related assays and is provided in small, sealed quantities for laboratory use.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Supplied in milligram-scale packages suitable for reference use.
  • Stable solid form for reliable storage and handling.
  • Recommended sealed storage to protect from moisture.
  • Suitable for LC-MS and PK method development.
Catalog No. 50-002-98422