Learn More
Medchemexpress LLC AMG-108 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9903211MG
Encompass_Preferred
AMG-108 is a humanized antibody expressed in CHO cells targeting IL-1R1/CD121a AMG-108 is composed of a huIgG2 heavy chain and a hu light chain with a predicted molecular weight (MW) of 144 62 kDa The isotype control for AMG-108 can refer to Human IgG2 kappa Isotype Control (HY-P99002)
Catalog No. 50-003-25779