Learn More
TARGETMOL CHEMICALS INC AMG-1694 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T142125MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. AMG-1694 is a potent glucokinase–glucokinase regulatory protein (GK-GKRP) disruptors and promotes the dissociation of the GK-GKRP complex with an IC50 of 7 nM indirectly increasing GK enzymatic activity. AMG-1694 normalizes blood glucose levels in several rodent models of diabetes and lowes blood glucose restricted to diabetic and not normoglycaemic animals[1]. AMG-1694 potently reverses the inhibitory effect of GKRP on GK activity and promotes GK translocation. purity: 97%
Catalog No. 50-258-1305