Learn More
Medchemexpress LLC HY-112703 5mg Medchemexpress, AMG 333 CAS:1416799-28-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127035MG
Encompass_Preferred
Medchemexpress, HY-112703 5mg AMG 333 CAS:1416799-28-4 AMG 333 is a potent and highly selective TRPM8 antagonist with an IC50 of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9147