Learn More
Medchemexpress LLC AMG-337 | 1173699-31-4 | 99.4% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1869610MG
Encompass_Preferred
AMG-337 is a potent, orally active, selective MET kinase inhibitor for preclinical research. It blocks MET phosphorylation and downstream signaling, reducing proliferation and inducing apoptosis in MET-amplified cancer models. Supplied as a well-characterized, high-purity solid for laboratory studies.
Catalog No. 50-002-94913