Learn More
Apexbio Technology LLC (±)-AMG 487 50mg
SDP
Supplier:  Apexbio Technology LLC B767150
Encompass_Preferred
racemic Baclofen -AMG 487 is a small-molecule antagonist targeting the chemokine receptor CXCR3 It is designed to inhibit ligand-induced CXCR3-mediated immune cell migration thereby modulating lymphocyte recruitment to inflammatory and metastatic sites racemic Baclofen -AMG 487 exerts its biological activity primarily through selective inhibition of CXCR3-mediated cell migration In in vitro studies racemic Baclofen -AMG 487 demonstrates inhibition of ligand-induced CXCR3-dependent cell movement Based on these pharmacological properties racemic Baclofen -AMG 487 holds research potential in inflammation and oncology studies
Catalog No. 50-000-92004