Learn More
Medchemexpress LLC HY-15319 50mg , AMG 487 CAS:473719-41-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1531950MG
Encompass_Preferred
HY-15319 50mg AMG 487 CAS: 473719-41-4 AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0904