Learn More
Medchemexpress LLC AMG 487 (S-enantiomer) | 473720-30-8 | C₃₂H₂₈F₃N₅O₄ | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15319B50MG
Encompass_Preferred
AMG 487 S-enantiomer is the S enantiomer of AMG 487 and functions as an antagonist of the chemokine receptor CXCR3. This chemical compound is provided as an off-white to light yellow solid with a purity of 99.94%.
  • Functions as an antagonist of the chemokine receptor CXCR3
  • High purity of 99.94%
  • Supplied as a solid
  • Soluble in DMSO
Catalog No. 50-003-72535