Learn More
Medchemexpress LLC AMG 487 (S-enantiomer) | 473720-30-8 | C32H28F3N5O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15319B10MM/1ML
Encompass_Preferred
AMG 487 (S-enantiomer) is the S enantiomer of AMG 487, a potent antagonist of the chemokine receptor CXCR3. This product is intended for research use only.
  • Acts as an antagonist of the chemokine receptor CXCR3
  • High purity (99.94%)
  • S-enantiomer form
  • Suitable for research applications
Catalog No. 50-003-72871