Learn More
Medchemexpress LLC AMG 487 (S-enantiomer) | 473720-30-8 | C32H28F3N5O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15319B5MG
Encompass_Preferred
AMG 487 S-enantiomer is the S enantiomer of AMG 487. It functions as an antagonist of the chemokine receptor CXCR3. This product is intended for research use only and is not sold to patients.
  • Antagonist of chemokine receptor CXCR3.
  • High purity (99.94%).
  • Solid appearance.
  • Off-white to light yellow color.
  • Recommended storage conditions for powder and in solvent.
Catalog No. 50-003-72696