Learn More
Apexbio Technology LLC AMG 510(Synonyms: Sotorasib, Lumakras, AMG510, KRAS G12C inhibitor, AMG-510), 10mg, CAS: 2296729-00-3.
SDP
Supplier:  Apexbio Technology LLC B839910
Encompass_Preferred
AMG 510 (CAS 2296729-00-3) is an orally bioavailable selective covalent inhibitor targeting KRAS G12C Its mechanism involves binding KRAS G12C mutant protein and stabilizing it in an inactive GDP-bound conformation By preventing nucleotide exchange AMG 510 suppresses downstream signaling pathways linked to cellular proliferation and survival resulting in tumor regression in preclinical KRAS G12C-driven cancer models It serves as an important tool for biomedical research focused on RAS-driven oncogenic signaling and targeted cancer therapeutics development
Catalog No. 50-000-59961