Learn More
Selleck Chemical LLC AMG 811-A2938-1MG-5
SDP
Supplier:  Selleck Chemical LLC A29381MG5
Encompass_Preferred
AMG 811 is a human monoclonal antibody (IgG1 isotype) that selectively targets human IFNgamma 7 It has potential to treat systemic lupus erythematosus (SLE) MW 144 36 KD
Catalog No. 50-313-4646