Learn More
Medchemexpress LLC HY-12661A 10mg , AMG PERK 44 CAS:1883548-84-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12661A10MG
Encompass_Preferred
HY-12661A 10mg AMG PERK 44 CAS: 1883548-84-2 AMG PERK 44 is an orally active and highly selective PERK inhibitor with an IC50 of 6 nM. AMG PERK 44 has 1000-fold and 160-fold selectivity over GCN2 (IC50=7300 nM) and B-Raf (IC50 >1000 nM), respectively. AMG PERK 44 induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9179