Learn More
Medchemexpress LLC Amg perk 44 | 1883548-84-2 | 99.67% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12661A10MM/1ML
Encompass_Preferred
AMG PERK 44 is an orally active and highly selective PERK inhibitor. It demonstrates significant selectivity over GCN2 and B-Raf, and is known to induce autophagy.
  • Orally active
  • Highly selective PERK inhibitor with an IC50 of 6 nM
  • Exhibits 1000-fold selectivity over GCN2 (IC50=7300 nM)
  • Exhibits 160-fold selectivity over B-Raf (IC50 >1000 nM)
  • Induces autophagy
Catalog No. 50-003-38880