Learn More
Ansh Labs AMH (Bovine) ELISA 96-wells, 50 L Plasma/Serum, Range 6 calibrators ranging 13.5-2240 pg/mL, LOD 11 pg/mL, Time 3.5 hours, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL114
Encompass
Anti-Mullerian Hormone AMH (Bovine) ELISA has a total 3.5 hours incubation, calibrators and range of 6, 13.5-2240 pg/mL, sensitivity of 11 pg/mL, sample of 50 µL Plasma. Reacts with Bovine, Caprine. Crossreactivity with related molecules is not significant. Highly published and validated for major breeds. In Bovine or other production animals, measuring AMH will predict response to ovarian stimulation and embryo yield, predict fertility in dairy cows: selection for culling, diagnose Granulosa-Theca Cell Tumors, test for the presence of functional testicular tissue. Ansh Labs, located just south of Houston Texas, is an FDA registered manufacturer of diagnostic test kits, antibodies, antigens, and proteins.
Catalog No. 50-194-3727