Learn More
Ansh Labs AMH ELISA 96-wells, 25 L Plasma/Serum, Range 6 calibrators ranging 0.084-14.2 ng/mL, LOD 23 pg/mL, Time 2.5 hours, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL105
Encompass
The AMH ELISA immunoassay uses standardized recombinant human AMH calibrators to ensure accuracy and reproducibility between assays and lots.  The Ansh Labs AMH ELISA uses unique monoclonal antibodies against specific linear epitopes on the associated dimers of AMH for specificity and consistent AMH detection.  The Anti-Mullerian Hormone ELISA is specific to human AMH (associated form) which detects the full length and enhanced biologically active associater forms of human AMH.  With an analytical measure range of 0.08 to 14.2 ng/mL and a sensitivity of 23 pg/mL, the AMH ELISA assay can measure AMH in studies ranging from menopause or compromised gonadal function up to PCOS.
Catalog No. 50-194-3721